82.48 GBP valid until 2025-12-31
Use promo code "24071" to get your promotional price.
Use promo code "24071" to get your promotional price.
Closure Material | HDPE |
---|---|
Capacity (Metric) | 15 mL |
Disposable | Single-use |
Material | Polypropylene |
No. per Case | 500/Cs. |
No. per Pack | 50 |
Packaging | Bulk |
Quantity | 50 Pk. |
Sterility | Sterile |
Green Features | Sustainable packaging |
RNase Free | Yes |
Dnase Free | Yes |
Autoclavable | Not Autoclavable |
Shape | Cylindrical |
Length (Metric) | 95 mm |
Bottom Shape | Conical |
Closure Type | Screw Cap |
Closure Included | Yes |
Writing Area | Yes |
Graduated | Yes |
Product Type | Centrifuge Tube |
Max. RCF | 10,500 x g |