Use promo code "25339" to get your promotional price.
Thermo Scientific Alfa Aesar 4-(Dimethylamino)pyridine, 99%, Thermo Scientific Chemicals
Use promo code "25339" to get your promotional price.
Quantity | 25 g |
---|
Chemical Identifiers
CAS | 1122-58-3 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.171 |
MDL Number | MFCD00006418 |
InChI Key | VHYFNPMBLIVWCW-UHFFFAOYSA-N |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
PubChem CID | 14284 |
IUPAC Name | N,N-dimethylpyridin-4-amine |
SMILES | CN(C)C1=CC=NC=C1 |
Specification
Chemical Name or Material | 4-Dimethylaminopyridine |
---|---|
CAS | 1122-58-3 |
Melting Point | 110°C to 113°C |
Boiling Point | 162°C (50 mmHg) |
Flash Point | 124°C (255°F) |
Assay Percent Range | 99% |
Molecular Formula | C7H10N2 |
MDL Number | MFCD00006418 |
Quantity | 25 g |
UN Number | UN2928 |
Beilstein | 110354 |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Synonym | 4-dimethylaminopyridine,dmap,4-dimethylamino pyridine,4-pyridinamine, n,n-dimethyl,n,n-dimethyl-4-pyridinamine,p-dimethylaminopyridine,pyridine, 4-dimethylamino,ccris 6176,dimethylpyridin-4-ylamine |
Solubility Information | Soluble in methanol,benzene,ethyl acetate,chloroform,methylene chloride,acetone,and acetic acid. Slightly soluble in ether,diisopropyl ether,cyclohexane,hexane,and water. |
InChI Key | VHYFNPMBLIVWCW-UHFFFAOYSA-N |
SMILES | CN(C)C1=CC=NC=C1 |
IUPAC Name | N,N-dimethylpyridin-4-amine |
Molecular Weight (g/mol) | 122.171 |
PubChem CID | 14284 |
Formula Weight | 122.17 |
Percent Purity | 99% |
Technical documents
[4-(Dimethylamino)pyridine]
[Danger]
[{code=ACUTE TOX. 2, sequence=1, description=Acute toxicity Category 2}, {code=ACUTE TOX. 3, sequence=2, description=Acute toxicity Category 3}, {code=AQUA CHRONIC 2, sequence=3, description=LONG-TERM AQUATIC HAZARD Chronic 2}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}, {code=SKIN IRRIT. 2, sequence=5, description=Skin corrosion/irritation Category 2}, {code=STOT SE 1, sequence=6, description=Specific target organ toxicity after single exposure Category 1}]
[{code=H301, sequence=1, description=Toxic if swallowed.}, {code=H310, sequence=2, description=Fatal in contact with skin.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H318, sequence=4, description=Causes serious eye damage.}, {code=H331, sequence=5, description=Toxic if inhaled.}, {code=H370, sequence=6, description=Causes damage to organs.}, {code=H411, sequence=7, description=Toxic to aquatic life with long lasting effects.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P308+P313, sequence=55, description=IF exposed or concerned: Get medical advice/attention.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : 4-(Dimethylamino)pyridine}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Product Identifier
- 4-(Dimethylamino)pyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H301- Toxic if swallowed.
- H310- Fatal in contact with skin.
- H315- Causes skin irritation.
- H318- Causes serious eye damage.
- H331- Toxic if inhaled.
- H370- Causes damage to organs.
- H411- Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
- P310- Immediately call a POISON CENTER/doctor/.
Supplemental Information
- MIXTURE LIST- Contains : 4-(Dimethylamino)pyridine