Thermo Scientific Alfa Aesar 3-Ethylcyclopentanone, 99%
CAS: 10264-55-8 | C7H12O | 112.172 g/mol
Overview
227.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 10264-55-8 Weight: 0.00500kg Transport: UN number : 1224 Chem class : 3 Pack group : III
Quantity 5 g
Disclaimers: RUO – Research Use Only
Product Code. 11414387
Quantity Price
1 £ 228.0 / 5g
Estimated Shipment
02-02-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 10264-55-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00045445
InChI Key XERALSLWOPMNRJ-UHFFFAOYSA-N
PubChem CID 139121
IUPAC Name 3-ethylcyclopentan-1-one
SMILES CCC1CCC(=O)C1
Specification
Chemical Name or Material 3-Ethylcyclopentanone
CAS 10264-55-8
Boiling Point 171°C
Flash Point 31°C (88°F)
Molecular Formula C7H12O
MDL Number MFCD00045445
Quantity 5 g
UN Number UN1224
Synonym 3-ethylcyclopentanone,cyclopentanone, 3-ethyl,3-ethyl cyclopentanone,cyclopentanone,3-ethyl,acmc-20an48,3-ethyl-cyclopentan-1-one
Synonym 3-ethylcyclopentanone,cyclopentanone, 3-ethyl,3-ethyl cyclopentanone,cyclopentanone,3-ethyl,acmc-20an48,3-ethyl-cyclopentan-1-one
Synonym 3-ethylcyclopentanone,cyclopentanone, 3-ethyl,3-ethyl cyclopentanone,cyclopentanone,3-ethyl,acmc-20an48,3-ethyl-cyclopentan-1-one
Synonym 3-ethylcyclopentanone,cyclopentanone, 3-ethyl,3-ethyl cyclopentanone,cyclopentanone,3-ethyl,acmc-20an48,3-ethyl-cyclopentan-1-one
Synonym 3-ethylcyclopentanone,cyclopentanone, 3-ethyl,3-ethyl cyclopentanone,cyclopentanone,3-ethyl,acmc-20an48,3-ethyl-cyclopentan-1-one
Synonym 3-ethylcyclopentanone,cyclopentanone, 3-ethyl,3-ethyl cyclopentanone,cyclopentanone,3-ethyl,acmc-20an48,3-ethyl-cyclopentan-1-one
Solubility Information Soluble in water.
InChI Key XERALSLWOPMNRJ-UHFFFAOYSA-N
SMILES CCC1CCC(=O)C1
IUPAC Name 3-ethylcyclopentan-1-one
Molecular Weight (g/mol) 112.172
PubChem CID 139121
Formula Weight 112.17
Percent Purity 99%
Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.