Use promo code "25339" to get your promotional price.
Thermo Scientific Alfa Aesar n-Heptane, 99%
Use promo code "25339" to get your promotional price.
Quantity | 500 mL |
---|
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Specification
Chemical Name or Material | n-Heptane |
---|---|
Melting Point | -91°C |
CAS | 142-82-5 |
Density | 0.684 |
Boiling Point | 97°C to 99°C |
Quantity | 500 mL |
Flash Point | −4°C (24°F) |
Odor | Petroleum-like |
Refractive Index | 1.3877 |
Molecular Formula | C7H16 |
MDL Number | MFCD00009544 |
UN Number | UN1206 |
Beilstein | 1730763 |
Merck Index | 14,4659 |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Synonym | n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish |
Solubility Information | Miscible with alcohol,chloroform,and ether. Insoluble in water. |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
SMILES | CCCCCCC |
IUPAC Name | heptane |
Molecular Weight (g/mol) | 100.21 |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
Formula Weight | 100.21 |
Percent Purity | 99% |
Technical documents
[n-Heptane]
[Danger]
[{code=AQUA CHRONIC 1, sequence=1, description=LONG-TERM AQUATIC HAZARD Chronic 1}, {code=AQUATIC ACUTE 1, sequence=2, description=ACUTE AQUATIC Acute 1}, {code=ASP. TOX. 1, sequence=3, description=Aspiration toxicity Category 1}, {code=FLAM. LIQ. 2, sequence=4, description=Flammable liquid Category 2}, {code=SKIN IRRIT. 2, sequence=5, description=Skin corrosion/irritation Category 2}, {code=STOT SE 3, sequence=6, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H304, sequence=2, description=May be fatal if swallowed and enters airways.}, {code=H315, sequence=3, description=Causes skin irritation.}, {code=H336, sequence=4, description=May cause drowsiness or dizziness.}, {code=H400, sequence=5, description=Very toxic to aquatic life.}, {code=H410, sequence=6, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P261, sequence=52, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P273, sequence=53, description=Avoid release to the environment.}, {code=P301+P310, sequence=54, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P302+P352, sequence=55, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P331, sequence=56, description=Do NOT induce vomiting.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : n-Heptane}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}, {code=GHS09, sequence=4, description=ENVIRONMENT}]
Product Identifier
- n-Heptane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H304- May be fatal if swallowed and enters airways.
- H315- Causes skin irritation.
- H336- May cause drowsiness or dizziness.
- H400- Very toxic to aquatic life.
- H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273- Avoid release to the environment.
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P331- Do NOT induce vomiting.
Supplemental Information
- MIXTURE LIST- Contains : n-Heptane
-
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular products
Buy Now