Use promo code "24128" to get your promotional price.
Thermo Scientific Alfa Aesar 1,2-Dimethoxyethane, 99+%, stab. with BHT
Use promo code "24128" to get your promotional price.
Quantity | 500 mL |
---|
Chemical Identifiers
CAS | 110-71-4 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.122 |
MDL Number | MFCD00008502 |
InChI Key | XTHFKEDIFFGKHM-UHFFFAOYSA-N |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
PubChem CID | 8071 |
ChEBI | CHEBI:42263 |
IUPAC Name | 1,2-dimethoxyethane |
SMILES | COCCOC |
Specification
Chemical Name or Material | 1,2-Dimethoxyethane |
---|---|
Name Note | stabilized with BHT |
CAS | 110-71-4 |
Melting Point | -69°C |
Density | 0.867 |
Boiling Point | 84°C to 86°C |
Flash Point | −6°C (21°F) |
Odor | Ether-like |
Molecular Formula | C4H10O2 |
Linear Formula | CH3OCH2CH2OCH3 |
Refractive Index | 1.38 |
MDL Number | MFCD00008502 |
Quantity | 500 mL |
UN Number | UN2252 |
Beilstein | 1209237 |
Merck Index | 14,3224 |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Synonym | monoglyme,ethylene glycol dimethyl ether,glyme,dimethyl cellosolve,egdme,ethane, 1,2-dimethoxy,dimethoxyethane,2,5-dioxahexane,glycol dimethyl ether,ethylene dimethyl ether |
Solubility Information | Miscible with water,methanol,ethanol,diethyl ether,acetone,tetrahydrofuran,benzene and toluene. |
InChI Key | XTHFKEDIFFGKHM-UHFFFAOYSA-N |
SMILES | COCCOC |
IUPAC Name | 1,2-dimethoxyethane |
Molecular Weight (g/mol) | 90.122 |
PubChem CID | 8071 |
ChEBI | CHEBI:42263 |
Formula Weight | 90.12 |
Percent Purity | ≥99% |
Technical documents
[1,2-dimethoxyethan]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=REPR. 1B, sequence=2, description=Reproductive toxicity Category 1B}, {code=ACUTE TOX. 4, sequence=3, description=Acute toxicity Category 4}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H332, sequence=2, description=Harmful if inhaled.}, {code=H360FD, sequence=3, description=May damage fertility. May damage the unborn child.}, {code=EUH019, sequence=91, description=May form explosive peroxides.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P201, sequence=52, description=Obtain special instructions before use.}, {code=P261, sequence=53, description=Avoid breathing dust/fume/gas/mist/vapours/spray.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P405, sequence=55, description=Store locked up.}, {code=P501, sequence=56, description=Dispose of contents/container to .}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- 1,2-dimethoxyethan
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Acute toxicity Category 4
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H332- Harmful if inhaled.
- H360FD- May damage fertility. May damage the unborn child.
- EUH019- May form explosive peroxides.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P201- Obtain special instructions before use.
- P261- Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405- Store locked up.
- P501- Dispose of contents/container to .