Use promo code "25339" to get your promotional price.
Thermo Scientific Alfa Aesar N,N-Dimethylacetamide, 99%
Use promo code "25339" to get your promotional price.
Quantity | 2500 mL |
---|
Chemical Identifiers
CAS | 127-19-5 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00008686 |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
PubChem CID | 31374 |
ChEBI | CHEBI:84254 |
IUPAC Name | N,N-dimethylacetamide |
SMILES | CN(C)C(C)=O |
Specification
Chemical Name or Material | N,N-Dimethylacetamide |
---|---|
CAS | 127-19-5 |
Melting Point | -20°C |
Density | 0.94 |
Boiling Point | 165°C to 166°C |
Flash Point | 70°C (158°F) |
Odor | Ammonia-like |
Molecular Formula | C4H9NO |
Linear Formula | CH3CON(CH3)2 |
Refractive Index | 1.4385 |
MDL Number | MFCD00008686 |
Quantity | 2500 mL |
Beilstein | 1737614 |
Merck Index | 14,3227 |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Synonym | dimethylacetamide,dmac,acetamide, n,n-dimethyl,acetdimethylamide,dimethyl acetamide,n,n-dimethyl acetamide,dimethylamide acetate,n,n-dimethylethanamide,dimethylacetone amide,acetyldimethylamine |
Solubility Information | Miscible with water,benzene,alcohol,acetone,ether,esters and ketones. |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
SMILES | CN(C)C(C)=O |
IUPAC Name | N,N-dimethylacetamide |
Molecular Weight (g/mol) | 87.12 |
PubChem CID | 31374 |
ChEBI | CHEBI:84254 |
Formula Weight | 87.12 |
Percent Purity | 99% |
Technical documents
[N,N-Dimethylacetamide]
[Danger]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=REPR. 1B, sequence=3, description=Reproductive toxicity Category 1B}]
[{code=H312, sequence=1, description=Harmful in contact with skin.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H332, sequence=3, description=Harmful if inhaled.}, {code=H360D, sequence=4, description=May damage the unborn child.}]
[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/soap.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P312, sequence=54, description=Call a POISON CENTER/doctor/if you feel unwell.}, {code=P337+P313, sequence=55, description=If eye irritation persists: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : N,N-Dimethylacetamide}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- N,N-Dimethylacetamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H312- Harmful in contact with skin.
- H319- Causes serious eye irritation.
- H332- Harmful if inhaled.
- H360D- May damage the unborn child.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352- IF ON SKIN: Wash with plenty of water/soap.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312- Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313- If eye irritation persists: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : N,N-Dimethylacetamide