Use promo code "25339" to get your promotional price.
1,3-Dimethyl-2-imidazolidinone, 98%
Use promo code "25339" to get your promotional price.
| Quantity | 100 g |
|---|
Chemical Identifiers
| CAS | 80-73-9 |
|---|---|
| Molecular Formula | C5H10N2O |
| Molecular Weight (g/mol) | 114.15 |
| MDL Number | MFCD00003188 |
| InChI Key | CYSGHNMQYZDMIA-UHFFFAOYSA-N |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| PubChem CID | 6661 |
| IUPAC Name | 1,3-dimethylimidazolidin-2-one |
| SMILES | CN1CCN(C)C1=O |
Specification
| Chemical Name or Material | 1,3-Dimethyl-2-imidazolidinone |
|---|---|
| CAS | 80-73-9 |
| Melting Point | 8°C |
| Density | 1.058 |
| Boiling Point | 224°C to 226°C |
| Flash Point | 93°C (199°F) |
| Molecular Formula | C5H10N2O |
| Refractive Index | 1.472 |
| MDL Number | MFCD00003188 |
| Quantity | 100 g |
| UN Number | UN2810 |
| Beilstein | 108808 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,3249 |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Synonym | 1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea |
| Solubility Information | Miscible with water. |
| InChI Key | CYSGHNMQYZDMIA-UHFFFAOYSA-N |
| SMILES | CN1CCN(C)C1=O |
| IUPAC Name | 1,3-dimethylimidazolidin-2-one |
| Molecular Weight (g/mol) | 114.15 |
| PubChem CID | 6661 |
| Formula Weight | 114.15 |
| Percent Purity | 98% |
Technical documents
[1,3-Dimethyl-2-imidazolidinone]
[Danger]
[{code=H302, sequence=1, description=Harmful if swallowed.}, {code=H312, sequence=2, description=Harmful in contact with skin.}, {code=H318, sequence=3, description=Causes serious eye damage.}, {code=H361, sequence=4, description=Suspected of damaging fertility or the unborn child.}]
[{code=P201, sequence=51, description=Obtain special instructions before use.}, {code=P202, sequence=52, description=Do not handle until all safety precautions have been read and understood.}, {code=P210, sequence=53, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P264, sequence=54, description=Wash face, hands and anyexposed skin thoroughly after handling.}, {code=P270, sequence=55, description=Do not eat, drink or smoke when using this product.}, {code=P280, sequence=56, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=57, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=58, description=Immediately call a POISON CENTER/doctor/.}, {code=P330, sequence=59, description=Rinse mouth.}, {code=P403+P235, sequence=60, description=Store in a well-ventilated place. Keep cool.}, {code=P501b, sequence=61, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: 1,3-Dimethylimidazolidin-2-one}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- 1,3-Dimethyl-2-imidazolidinone
Signal Word
- Danger
Hazard Statement
- H302- Harmful if swallowed.
- H312- Harmful in contact with skin.
- H318- Causes serious eye damage.
- H361- Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201- Obtain special instructions before use.
- P202- Do not handle until all safety precautions have been read and understood.
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264- Wash face, hands and anyexposed skin thoroughly after handling.
- P270- Do not eat, drink or smoke when using this product.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
- P330- Rinse mouth.
- P403+P235- Store in a well-ventilated place. Keep cool.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contain: 1,3-Dimethylimidazolidin-2-one