Code:
50
Gibco™ RPMI 1640 Medium, powder, HEPES
RPMI 1640 Medium, powder, HEPES
Additional Details:
Weight:
0.01100kg
Specification
Form | Powder |
---|---|
Product Type | RPMI 1640 Medium |
With Additives | Glutamine,HEPES,Phenol Red |
With Additives | Glutamine,HEPES,Phenol Red |
With Additives | Glutamine,HEPES,Phenol Red |
Without Additives | No Sodium Bicarbonate,No Sodium Pyruvate |
Without Additives | No Sodium Bicarbonate,No Sodium Pyruvate |
For Use With (Application) | Mammalian cell culture |
Quantity | 10 L |
Certifications/Compliance | ISO 13485 standards, FDA |
Regulatory Status | For in-vitro diagnostic use |
Content And Storage | 2 to 8°C |
Description | RPMI 1640 Medium, 10L, Powdered Form, With L-glutamine, Phenol Red, HEPES, 36 Months Shelf Life, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |