355.28 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Column Format | Guard Cartridge, 4 Pk. |
---|---|
Diameter (Metric) | 4 mm |
Length (Metric) | 10 mm |
Max. Pressure | 5800 psi (400 bar) |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
USP Type | L7 |
Stationary Phase | C8 |
Product Line | Hypersil GOLD |
Packing Material | Silica |
Carbon Load | 8% |
Column Type | Reversed Phase |
Endcapped | Yes |
Phase | Reversed Phase |