179.83 GBP valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Use promo code "24147" to get your promotional price.
Cell Line | CHO |
---|---|
Concentration | 1 X |
Form | Liquid |
Product Type | Ham's F-12 Nutrient Mix |
With Additives | Glutamine,Phenol Red,Sodium Pyruvate |
With Additives | Glutamine,Phenol Red,Sodium Pyruvate |
With Additives | Glutamine,Phenol Red,Sodium Pyruvate |
For Use With (Application) | Mammalian cell culture |
Quantity | 10 x 500 mL |
Certifications/Compliance | ISO 13485 standards, FDA |
pH | 7.1 to 7.5 |
Content And Storage | 2 to 8°C |
Description | Ham's F-12 Nutrient Mix, 10 x 500mL, 1 X Concentration, Liquid, 7.1 to 7.5 pH, 12 Months Shelf Life, 280 to 310 mOsm/kg Osmolality, With Sodium Pyruvate, L-glutamine, Phenol Red, 2 to 8°C Storage, For Mammalian Cell Culture |