Code:
66
Falcon™ Standard Tissue Culture Dishes
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Additional Details:
Weight:
4.68000kg
Specification
Bottom Shape | Flat |
---|---|
Capacity (Metric) | 7 mL (Well) |
Color | Clear |
Culture Area | 21.29 cm2 |
Material | Polystyrene |
Shape | Round |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Type | Cell Culture Petri Dish |
Volume (Metric) Working | 6 to 7 mL |
No. per Case | 500/Cs. |
No. per Pack | 20/Pk. |
Height (Metric) | 15 mm |
Diameter (Metric) | 60 mm |