Code:
50
Gibco™ McCoy's 5A (Modified) Medium
McCoy's 5A (Modified) Medium
Additional Details:
Weight:
0.50000kg
Specification
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | McCoy's 5A (Modified) Medium |
With Additives | High Glucose,Glutamine,Phenol Red,Bacto-peptone |
With Additives | High Glucose,Glutamine,Phenol Red,Bacto-peptone |
With Additives | High Glucose,Glutamine,Phenol Red,Bacto-peptone |
With Additives | High Glucose,Glutamine,Phenol Red,Bacto-peptone |
Without Additives | No HEPES,No Sodium Pyruvate |
Without Additives | No HEPES,No Sodium Pyruvate |
For Use With (Application) | Mammalian cell culture |
Quantity | 500 mL |
Certifications/Compliance | ISO 13485 standards, FDA |
pH | 6.9 to 7.1 |
Regulatory Status | For in-vitro diagnostic use |
Content And Storage | 2 to 8°C |
Description | McCoy's 5A (Modified) Medium, 500mL, 1 X Concentration, Liquid Form, With High Glucose, L-glutamine, Bacto-peptone, Phenol Red, 6.9 to 7.1 pH, 12 Months Shelf Life, 280 to 320mOsm/kg Osmolality, Used for Mammalian Cell Culture |