10.64 GBP valid until 2025-12-31
Use promo code "24335" to get your promotional price.
Use promo code "24335" to get your promotional price.
Chelators | EDTA |
---|---|
Content And Storage |
Storage conditions: -5°C to -20°C Shelf life: 24 months from date of manufacture |
Cell Type | Mammalian Cells |
Classification | Animal Origin |
Concentration | 1 X |
Form | Liquid |
Product Type | Cell Culture Dissociation Reagent |
Sterility | Sterile-filtered |
Sterilization Method | Sterile-filtered |
With Additives | Phenol Red,EDTA |
With Additives | Phenol Red,EDTA |
Osmolality | 270 - 310 mOsm/kg |
pH | 7.1 to 8 |
Product Line | Gibco™ |
Quantity | 100 mL |
Shipping Condition | Wet ice |
Source | Animal Origin |
Tested For | In vitro Bioassay |