Thermo Scientific™ Heratherm™ Advanced Protocol Ovens, 230VAC 60Hz
Carry out multiple applications with this advanced protocol oven, which features temperature uniformity and low energy-consumption.
Overview
1590.65 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: Weight: 0.10000kg
Disclaimers: Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 11606430
Quantity Price
1 £ 2381.73 / Each
Estimated Shipment
12-09-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Accessories and Spare Parts
  • Special Offers
Description and Specification
Specification
Controller Type Programmable Microprocessor control with vacuum fluorescent display
Description 51028142
Capacity (Metric) 61 L
Temperature (Metric) 330°C
Type Gravity Convection
Material Coated Steel
No. of Shelves 2 supplied/13 max.
Plug Type Country-specific
Temperature Stability ±2.5°C
Dimensions (D x W x H) Interior 15.3 x 12.9 x 18.9 in. (38.9 x 32.8 x 48 cm)
Dimensions (L x W x H) 22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English) 22.2 in.
Length (Metric) 56.5 cm
Width (English) 20.9 in.
Width (English) Interior 12.9 in.
Width (Metric) 53 cm
Width (Metric) Interior 32.8 cm
Height (English) 28.3 in.
Height (English) Interior 18.9 in.
Height (Metric) 72 cm
Height (Metric) Interior 48 cm
Depth (English) Interior 15.3 in.
Depth (Metric) Interior 38.9 cm
Electrical Requirements 230 V 50/60 Hz
Voltage 230 V
Wattage 1800 w
Amperage 7.9 A
Frequency 50/60 Hz
Heratherm Advanced Protocol Ovens provide a choice of gravity or mechanical convection. Gravity convection units are designed to protect delicate samples while offering temperature uniformity of ±2.9°C or better, and temperature stability of at least ±0.3°C at 150°C. Mechanical convection models provide temperature uniformity of ±3.0°C or better, and temperature stability of at least ±0.2°C at 150°C.
  • Broad temperature range—up to 330°C
  • Adjustable fan speed for application-related airflow in mechanical convection models (e.g. drying of powders)
  • High speed for expedited drying and heating, quick recovery, and optimal temperature stability and uniformity
  • Programmable controller for temperature ramps and dwells
  • Up to 10 programs can be saved, each with 10 discrete steps
  • Features electronically controlled fan speed and damper position
  • Programs can be repeated automatically
  • Access port allows the introduction of sensors for independent data monitoring
  • Simple calibration routine ensures temperature accuracy over time
  • Boost function enables rapid heating up—no need to operate the oven 24/7 (not available in large capacity units)
Ordering Information:
Availability may vary by country.