Code:
88
Thermo Scientific™ DreamTaq PCR Master Mixes (2X)
Thermo Scientific DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs.
Additional Details:
Weight:
2.44490kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Content And Storage |
• 2X DreamTaq Green PCR Master Mix, 20 x 1.25 mL • Nuclease-free water, 20 x 1.25 mL Store at -20°C. |
---|---|
GC-Rich PCR Performance | Low |
Polymerase | DreamTaq DNA Polymerase |
Reaction Speed | Standard |
Quantity | 20 x 1.25 mL |
Shipping Condition | Dry Ice |
For Use With (Application) | Standard PCR |
Concentration | 2X |
Fidelity (vs. Taq) | 1X |
Hot Start | No |
No. of Reactions | 1000 Reactions |
Overhang | 3'-A |
Reaction Format | SuperMix or Master Mix |
Color | Green |