Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl
Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.
Overview
Code: PN
Additional Details:
Additional Details: CAS Number: 151-21-3 Weight: 0.10000kg Transport: UN number : 2926 Chem class : 4.1 Pack group : II
Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Product Code. 11821475
Quantity Price
1 £ 105.0 / 100g
In stock, ready to ship: 3
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
Description and Specification
Specification
Color White
Product Type Detergent
Solubility 10% (aq, w/v) solution must be clear, colorless to slightly yellow
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Powder
Reagent Type Detergent (Pure)
Quantity 100 g
This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).
Highlights:
  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
  • Common component of cell lysis buffers
Detergent properties (pure C12):
  • Molecular Weight: 288.5
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 62
  • Micelle Molecular Weight: 18,000
  • Critical Micelle Concentration (CMC): 6 to 8mM (0.1728 to 0.2304%, w/v)
  • Cloud Point: greater than 100°C
  • Dialyzable: No
Storage Requirements
  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
  • Should be stored between the following temperatures: 20°C to 25°C
Alternative Products
Safety and Handling
[sodium dodecyl sulphate]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=AQUATIC ACUTE 1, sequence=3, description=ACUTE AQUATIC Acute 1}, {code=EYE DAM. 2, sequence=4, description=Serious eye damage/eye irritation Category 2}, {code=FLAM. SOL. 2, sequence=5, description=Flammable solid Category 2}, {code=SKIN IRRIT. 2, sequence=6, description=Skin corrosion/irritation Category 2}, {code=STOT SE 3, sequence=7, description=Specific target organ toxicity Category 3}]
[{code=H228, sequence=1, description=Flammable solid.}, {code=H302, sequence=2, description=Harmful if swallowed.}, {code=H311, sequence=3, description=Toxic in contact with skin.}, {code=H315, sequence=4, description=Causes skin irritation.}, {code=H319, sequence=5, description=Causes serious eye irritation.}, {code=H335, sequence=6, description=May cause respiratory irritation.}, {code=H400, sequence=7, description=Very toxic to aquatic life.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P264, sequence=52, description=Wash thoroughly after handling.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : sodium dodecyl sulphate}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS09, sequence=3, description=ENVIRONMENT}]
Product Identifier
  • sodium dodecyl sulphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228- Flammable solid.
  • H302- Harmful if swallowed.
  • H311- Toxic in contact with skin.
  • H315- Causes skin irritation.
  • H319- Causes serious eye irritation.
  • H335- May cause respiratory irritation.
  • H400- Very toxic to aquatic life.
Precautionary Statement
  • P260- Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264- Wash thoroughly after handling.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental Information
  • MIXTURE LIST- Contains : sodium dodecyl sulphate