203.30 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.
Use promo code "24163" to get your promotional price.
Description | Septa |
---|---|
Color | Dark Blue,White |
Color | Dark Blue,White |
Diameter (Metric) Outer | 8 mm |
Material | PTFE,Silicone |
Material | PTFE,Silicone |
Quantity | 100 |
Thickness (Metric) | 1.3 mm |