154.94 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Use promo code "25721" to get your promotional price.
Diameter (English) Outside | 0.7 in. |
---|---|
Capacity (Metric) | 15 mL |
Material | Polypropylene |
Includes | Black graduations |
No. per Case | 500 |
No. per Pack | 50 |
Packaging | Racked |
Tolerance | The recommended working temperature range for polypropylene and PET is the same (0 - 40°C) |
Sterility | Sterile |
Shape | Conical |
Length (Metric) | 119 mm |
Capacity (English) | 0.50 oz. |
Bottom Shape | Conical |
Closure Type | Flat Cap |
Writing Area | White Marking Spot |
Diameter (Metric) | 17 mm |
Graduations | 0.5 mL |
Product Type | Centrifuge Tube |
Max. RCF | 12,000 x g |