71.36 GBP valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Use promo code "24108" to get your promotional price.
No. per Case | 100 |
---|---|
No. per Pack | 5 |
Product Type | Cell Culture Flask |
Surface Coating | Cell Culture Treated |
Sterility | Sterile |
Bottom Shape | Flat |
Closure Type | Vented |
Culture Area | 75 cm2 |
Surface Treatment | Cell Culture-treated |
Volume (Metric) Working | 25 mL |