330.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Use promo code "25977" to get your promotional price.
Description | 919 Series, Clear |
---|---|
Volume (Metric) | 250 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Clear |
Autoclavable | Autoclavable |
Certifications/Compliance | ISO 9001 and ISO 13485 |
Clean Claims | Free of RNase, DNase, DNA, ATP, and endotoxin |
Format | Rack |
Tip Style | Standard |
Packaging | 96 tips/rack, 10 racks/pack, 5 packs/case |
Quantity | 4800 tips |
For Use With (Equipment) | Beckman Coulter Biomek FX,Beckman Coulter Biomek NX,Beckman Coulter Biomek 3000,Beckman Coulter Multimek AP96 |
For Use With (Equipment) | Beckman Coulter Biomek FX,Beckman Coulter Biomek NX,Beckman Coulter Biomek 3000,Beckman Coulter Multimek AP96 |
For Use With (Equipment) | Beckman Coulter Biomek FX,Beckman Coulter Biomek NX,Beckman Coulter Biomek 3000,Beckman Coulter Multimek AP96 |
For Use With (Equipment) | Beckman Coulter Biomek FX,Beckman Coulter Biomek NX,Beckman Coulter Biomek 3000,Beckman Coulter Multimek AP96 |
Product Line | Beckman |
Fit for Purpose | Certified Quality |