167.99 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Certifications/Compliance | Produced and tested in accordance with the EN ISO 24998:2008 Petri dish standard which includes stringent dimensional controls. |
---|---|
Description | Petri dish, 90mm |
Material | Polystyrene |
Shape | Round |
Sterility | Sterile |
Surface Treatment | Non-treated |
Type | Petri Dish |
No. per Case | 500 |
No. per Pack | 20 |
Vent | 1 |
Height (Metric) | 15.9 mm |
Height (English) | 15.9 in. |
Diameter (Metric) | 90 mm |