Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Sodium tert-butoxide, 99%, pure
Use promo code "25339" to get your promotional price.
Quantity | 2.5 kg |
---|---|
Packaging | Plastic Drum |
Chemical Identifiers
CAS | 865-48-5 |
---|---|
Molecular Formula | C4H9NaO |
Molecular Weight (g/mol) | 96.1 |
MDL Number | MFCD00040575 |
InChI Key | MFRIHAYPQRLWNB-UHFFFAOYSA-N |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
PubChem CID | 23676156 |
IUPAC Name | sodium;2-methylpropan-2-olate |
SMILES | CC(C)(C)[O-].[Na+] |
Specification
Chemical Name or Material | Sodium tert-butoxide |
---|---|
CAS | 865-48-5 |
Melting Point | 180.0°C |
Flash Point | 12°C |
Assay Percent Range | 99% |
Packaging | Plastic Drum |
Molecular Formula | C4H9NaO |
Linear Formula | NaOC(CH3)3 |
MDL Number | MFCD00040575 |
Quantity | 2.5 kg |
Fieser | 16,55 |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Synonym | sodium tert-butoxide,sodium 2-methylpropan-2-olate,sodium t-butoxide,sodium-t-butoxide,t-butoxy sodium,tert-butoxysodium,naotbu,tbuona,2-propanol, 2-methyl-, sodium salt,sodium-tert-butoxide |
Solubility Information | Solubility in water: reacts. |
InChI Key | MFRIHAYPQRLWNB-UHFFFAOYSA-N |
SMILES | CC(C)(C)[O-].[Na+] |
IUPAC Name | sodium;2-methylpropan-2-olate |
Molecular Weight (g/mol) | 96.1 |
PubChem CID | 23676156 |
Formula Weight | 96.1 |
Percent Purity | 99% |
Grade | Pure |
Technical documents
[Sodium tert-butoxide]
[Danger]
[{code=EYE DAM. 1, sequence=1, description=Serious eye damage/eye irritation Category 1}, {code=FLAM. SOL. 1, sequence=2, description=Flammable solid Category 1}, {code=SELF-HEAT. 1, sequence=3, description=Self-heating substance Category 1}, {code=SKIN CORR. 1B, sequence=4, description=Skin corrosion/irritation Category 1 B}]
[{code=H318, sequence=1, description=Causes serious eye damage.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H251, sequence=3, description=Self-heating: may catch fire.}, {code=H228, sequence=4, description=Flammable solid.}, {code=EUH014, sequence=91, description=Reacts violently with water.}]
[{code=P210, sequence=50, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=53, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=54, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}]
Product Identifier
- Sodium tert-butoxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318- Causes serious eye damage.
- H314- Causes severe skin burns and eye damage.
- H251- Self-heating: may catch fire.
- H228- Flammable solid.
- EUH014- Reacts violently with water.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.