Chemical Name or Material |
Glycerol
|
CAS |
56-81-5
|
Decomposition Information |
>290°C
|
Melting Point |
18.0°C
|
Density |
1.2610g/mL
|
Boiling Point |
290.0°C
|
Flash Point |
160°C
|
Ignition Residue |
0.005% max.
|
Infrared Spectrum |
Authentic
|
Assay Percent Range |
99.5% min. (GC)
|
Packaging |
Plastic drum
|
Molecular Formula |
C3H8O3
|
Linear Formula |
HOCH2CH(OH)CH2OH
|
Refractive Index |
1.4716
|
MDL Number |
MFCD00004722
|
Quantity |
10 L
|
Beilstein |
01, 502
|
Merck Index |
15, 4520
|
Specific Gravity |
1.261
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Synonym |
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
|
Solubility Information |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
|
InChI Key |
PEDCQBHIVMGVHV-UHFFFAOYSA-N
|
SMILES |
OCC(O)CO
|
IUPAC Name |
propane-1,2,3-triol
|
Molecular Weight (g/mol) |
92.09
|
PubChem CID |
753
|
ChEBI |
CHEBI:17754
|
Viscosity |
1069 mPa.s (20°C)
|
Formula Weight |
92.09
|
Vapor Pressure |
0.003mbar at 50°C
|
Percent Purity |
99.6%
|
Grade |
ACS Reagent
|
Physical Form |
Liquid
|