Chemical Name or Material |
Sulfuric acid
|
CAS |
7664-93-9
|
Melting Point |
10.0°C
|
Boiling Point |
290.0°C
|
Density |
1.8300g/mL
|
Heavy Metals (as Pb) |
1ppm max.
|
Packaging |
Plastic drum
|
Residue after Ignition |
0.0005% max.
|
Specific Gravity |
1.83
|
Assay Percent Range |
95%
|
Quantity |
10 L
|
Molecular Formula |
H2O4S
|
Viscosity |
21 mPa.s (25°C)
|
Linear Formula |
H2SO4
|
Physical Form |
Liquid Free From Suspended or Insoluble Matter
|
MDL Number |
MFCD00064589
|
Fieser |
01,470; 05,633; 06,558; 07,347; 09,441
|
Merck Index |
15, 9104
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Synonym |
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
|
Solubility Information |
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
|
InChI Key |
QAOWNCQODCNURD-UHFFFAOYSA-N
|
SMILES |
OS(O)(=O)=O
|
IUPAC Name |
sulfuric acid
|
Molecular Weight (g/mol) |
98.07
|
PubChem CID |
1118
|
ChEBI |
CHEBI:26836
|
Formula Weight |
98.08
|
Percent Purity |
95.0 to 98.0%
|
Grade |
ACS Reagent
|