Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Bar-Coded Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Additional Details:
Weight:
7.48427kg
Specification
Format | Standard |
---|---|
Lid | With Lid |
Culture Area | 0.32 cm2 |
Array | 8 x 12 |
Color | Black,Clear Bottom |
Color | Black,Clear Bottom |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Well Shape | Flat Bottom (F-well) |
No. per Case | 100/Cs. |
No. per Pack | 20/Pk. |