Use promo code "27189" to get your promotional price.
Invitrogen™ NuPAGE™ Bis-Tris Mini Protein Gels, 4–12%, 1.0–1.5 mm
Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity.
78.54 GBP valid until 2025-12-31
Use promo code "27189" to get your promotional price.
Use promo code "27189" to get your promotional price.
Code:
50
Additional Details:
Weight:
0.00454kg
Includes:
One box containing 10 gels
Specification
For Use With (Equipment) | Mini Gel Tank,XCell SureLock Mini-Cell |
---|---|
For Use With (Equipment) | Mini Gel Tank,XCell SureLock Mini-Cell |
Gel Percentage | 4 to 12% |
Gel Size | Mini |
GelThickness | 1.5 mm |
Gel Type | Bis-Tris |
Recommended Applications | Denaturing |
Sample Loading Volume | Up to 25 μL |
Separation Range | 3.5 to 260 kDa |
Separation Type | Denaturing |
Wells | 15-well |
Mode of Separation | Molecular Weight |
Product Line | NuPAGE™ |
Quantity | 10 gels (1 box) |
Shipping Condition | Room Temperature |
Storage Requirements | Store at 4–25°C. Do not freeze. |
Length (Metric) | 8 cm |
Width (Metric) | 8 cm |