1601.95 GBP valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Use promo code "24069" to get your promotional price.
Capacity (English) | 2.3 cu. ft. |
---|---|
Capacity (English) Shelf | 55 lb. |
Controller Type | Microprocessor control with vacuum fluorescent display |
Capacity (Metric) | 66 L |
Convection Technology | Dual convection |
No. of Shelves | 2 supplied/ 13 max. |
Plug Type | Plug type provided based on country |
Temperature Range (Metric) | Ambient +5° to 105°C |
Type | Microbiological Incubator |
Temperature Stability | ±0.1°C |
Temperature Uniformity | ±0.6°C |
Timer | weekly / real time / hour |
Dimensions (D x W x H) Interior | 14.5 x 13.9 x 20 in. (36.8 x 35.4 x 50.8 cm) |
Dimensions (L x W x H) | 22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm) |
Length (English) | 22.2 in. |
Length (Metric) | 56.5 cm |
Width (English) | 20.9 in. |
Width (English) Interior | 13.9 in. |
Width (Metric) | 53 cm |
Width (Metric) Interior | 35.4 cm |
Height (English) | 28.3 in. |
Height (English) Interior | 20 in. |
Height (Metric) | 72 cm |
Height (Metric) Interior | 50.8 cm |
Depth (English) Interior | 14.5 in. |
Depth (Metric) Interior | 36.8 cm |
Weight (English) | 99 lb. |
Weight (Metric) | 45 kg |
Electrical Requirements | 230 V, 50/60 Hz |
Voltage | 230 V |
Wattage | 850 w |
Amperage | 3.7 A |
Frequency | 50/60 Hz |