Code:
A0
Fisherbrand™ 0.5mL Flat-Cap PCR Tubes
With integral snap shut cap
Additional Details:
Weight:
0.40000kg
Specification
Certifications/Compliance | RNase, Dnase and human DNA free |
---|---|
Color | Green |
Volume (Metric) | 0.5 mL |
Quantity | 1000 Case |
For Use With (Equipment) | 0.5mL thermal cycler blocks and can be used in PCR and sample storage applications |
Material | Polypropylene |
Closure Type | Flat Cap |
Format | Tube |
Product Type | Thin Wall PCR Tube |