80.93 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
Closure Material | Polypropylene |
---|---|
Autoclavable | Autoclavable |
Capacity (Metric) | 1.5 mL |
Color | Natural |
Material | Polypropylene |
Includes | O-ring |
Sterility | Sterile |
Closure Type | Screw Cap |
Bottom Shape | Conical |
Max. RCF | 30,000 x g |