15.64 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Additive | No Sodium Pyruvate |
---|---|
Classification | Animal Origin-Free |
Color | White |
Content And Storage |
Storage conditions: 15-30° C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
Dilution | 1 X |
Product Line | Gibco™ |
Product Type | Phosphate Buffered Saline (PBS) |
Quantity | 500 mL |
Solution Type | Phosphate Buffered Saline |
For Use With (Application) | Mammalian Cell Culture |
Osmolality | 280 - 320 mOsm/kg |
pH | 7.2 |
Phenol Red Indicator | No Phenol Red |
Shipping Condition | Room Temperature |
Sterility | Sterile-filtered |
Form | Liquid |
Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |