71.30 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Use promo code "24107" to get your promotional price.
Barcode | Includes Barcode |
---|---|
Certifications/Compliance | Certified RNase/DNase-free |
Packaging | 25 Plate |
Product Line | Armadillo |
Color | Clear |
Product Type | PCR Microplate |
For Use With (Equipment) | Real-Time PCR Systems or Thermal Cyclers |
Skirt Style | Skirted |
Material | Polycarbonate,Polypropylene |
Material | Polycarbonate,Polypropylene |
Volume (Metric) Well | 0.2 mL |
No. of Wells | 96 |
Volume (Metric) Working | 0.1 mL |
Quantity | 25 plates |
Reaction Speed | Low profile |
Well Color | Clear |