36.17 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Use promo code "24156" to get your promotional price.
ANSI Abrasion Rating | ANSI 5 |
---|---|
Certifications/Compliance | FDA 21 CFR 177.2600 |
Certifications/Compliance (Europe) | EN 1149 |
Chlorinated | Yes |
Applications | Industrial |
Color | Green |
Finger Thickness (English) | 0.0169 in. |
Finger Thickness (Metric) | 0.43 mm |
Latex Free | Yes |
Material | Nitrile |
Palm Thickness (English) | 0.0169 in. |
Palm Thickness (Metric) | 0.43 mm |
Product Type | Chemical-resistant Gloves |
Size | 8 |
Thickness (English) | 15 mil |
Thickness (Metric) | 0.38 mm |
EN388 Standard | EN 388:4101X, EN 388:4101 |
EN 421 | No |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | JKLOPT |
EN ISO 374 - 5 | Bacteria, Fungi only |
For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
Glove Cuff Style | Gauntlet |
Grip Design | Reversed Lozenge |
Liner Material | Cotton Flock |
Lining | Lined |
Model | 37-676 8 117722 |
Series | AlphaTec™ Solvex™ 37-676 |
Length (English) | 13 in. |
Length (Metric) | 33 cm |