Thermo Scientific™ MacConkey Agar without Salt (Dehydrated)
Detect and enumerate coliform organisms in clinical samples with this medium.
Overview
495.97 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: 64
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 12771540
Quantity Price
1 £ 514.0 / 2.50kg
Estimated Shipment
05-06-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Description MacConkey Agar w/o Salt (Dehydrated)
Form Powder
Product Type Agar
Quantity 2.5 kg
Yield for 53.2L medium
MacConkey Agar without Salt has the same formulation as MacConkey Agar except that it does not contain added salt and, therefore, provides a ‘low electrolyte medium’ on which most Proteus species do not spread. The medium has found particular favour for use in the examination of urine samples so that overgrowth of other organisms is prevented.