Code:
50
Memmert™ Forced Convection Standard Incubator with Single Display and Touchscreen, 53 L, Stainless Steel
Temperature Range: 10°C to 80°C
Additional Details:
Weight:
57.00000kg
Specification
Certifications/Compliance | CE |
---|---|
Controller Type | Multi-functional digital PID-microprocessor controller |
Description | Incubator |
Capacity (Metric) | 53 L |
Chamber Material | Stainless Steel |
Material | Stainless Steel |
Plug Type | UK |
Temperature Range (Metric) | 10°C to 80°C |
Type | Standard Incubator |
Display | TFT Color |
Model | IF55 |
No. of Grids | 4 |
Temperature Accuracy | 0.1°K |
Temperature Sensor | Pt100 |
Dimensions (D x W x H) Interior | 330 x 400 x 400 mm |
Dimensions (L x W x H) | 784 H x 585 W x 514 D mm |
Width (Metric) | 585 mm |
Width (Metric) Interior | 400 mm |
Height (Metric) | 784 mm |
Height (Metric) Interior | 400 mm |
Depth (Metric) Interior | 330 mm |
Shipping Weight (Metric) | 78 kg |
Weight (Metric) | 57 kg |
Electrical Requirements | 115/230 V 50/60 Hz |
Voltage | 115/230 V |
Wattage | 850/1 kW |
Frequency | 50/60 Hz |
-
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
Buy Now