1681.24 GBP valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Use promo code "24075" to get your promotional price.
Certifications/Compliance | CE |
---|---|
Controller Type | Multi-functional digital PID-microprocessor controller |
Description | Incubator |
Capacity (Metric) | 74 L |
Chamber Material | Stainless Steel |
Material | Stainless Steel |
Plug Type | UK |
Temperature Range (Metric) | 10°C to 80°C |
Type | Standard Incubator |
Display | TFT Color |
Model | IF75 |
No. of Grids | 6 |
Temperature Accuracy | 0.1°K |
Temperature Sensor | Pt100 |
Dimensions (D x W x H) Interior | 330 x 400 x 560 mm |
Dimensions (L x W x H) | 944 H x 585 W x 514 D mm |
Width (Metric) | 585 mm |
Width (Metric) Interior | 400 mm |
Height (Metric) | 944 mm |
Height (Metric) Interior | 560 mm |
Depth (Metric) Interior | 330 mm |
Shipping Weight (Metric) | 87 kg |
Weight (Metric) | 66 kg |
Electrical Requirements | 115/230 V 50/60 Hz |
Voltage | 115/230 V |
Wattage | 1.1/1.2 kW |
Frequency | 50/60 Hz |