Trajan Scientific and Medical™ BP5MS GC Capillary Column
SGE GC column BP5MS is an optimized silphenylene column for general purpose MS analysis
Overview
Code: 16
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12914005
Quantity Price
1 £ 441.87 / Each
Estimated Shipment
10-07-2026
Specification
Max. Temperature 350°C
Type GC Capillary column
Diameter (Metric) Inner 0.25 mm
For Use With (Application) 5% GC-MS Analysis
Temperature Range -40°C to +330/350°C
Film Thickness 0.25 μm
Length (Metric) 30 m
Material 5% Phenyl Polysilphenylene-siloxane
  • Perfect for 5% GCMS analysis
  • Optimized silphenylene content for general purpose MS analyses
Alternative Products