6.34 GBP valid until 2025-06-21
Use promo code "24097" to get your promotional price.
Use promo code "24097" to get your promotional price.
Closure Color | Blue |
---|---|
Material (Liner) | Silicone/PTFE |
Color | Amber |
Diameter (Metric) Outer | 11.6 mm |
Material | Glass |
Pre-slit | No |
Volume (Metric) | 1.85 mL |
No. per Pack | 100 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Height (Metric) | 32 mm |
Type | Hydrolytic Glass (Class I) |
Closure Size | 9-425 |
Marking Spot | Yes |