41.61 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
UserName
Product Code. 12930891
Quantity | Price |
---|---|
1 | £ 71.3 / Pack of 100 |
Closure Color | Blue |
---|---|
Material (Liner) | Silicone/PTFE |
Color | Amber |
Diameter (Metric) Outer | 11.6 mm |
Material | Hydrolytic Glass (Class I) |
Volume (Metric) | 1.85 mL |
No. per Pack | 100 |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Height (Metric) | 32 mm |
Closure Size | 9-425 |
Marking Spot | Yes |
There was an issue loading the e-mail form