119.66 GBP valid until 2025-06-08
Use promo code "24131" to get your promotional price.
Use promo code "24131" to get your promotional price.
Product Type | General Purpose Lab Coat |
---|---|
Size | Medium |
Gender | Unisex |
Material | Polypropylene |
Color | White |
Closure Type | Zipper |
Cuff Style | Elastic |
Disposable | Single-use |
Resistance | Non-hazardous Liquids,Dust |
Resistance | Non-hazardous Liquids,Dust |
Seam Type | Serged |
Size (English) Chest | 32 to 36 in. |
Size (Metric) Chest | 81 to 91 cm |
Certifications/Compliance (Europe) | CE Category 1 |
For Use With (Application) | Non-hazardous liquids and dust |
Includes | Internal pocket on the wearers left hand side (measuring 13 H x 12cm W) |
Packaging Type | Individually Packaged |