Fisherbrand™ 9 mm Short Thread Vial Kit
Includes 100 vials and 100 caps with pre-assembled septa.
Overview
36.90 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Code: 17
Additional Details:
Additional Details: Weight: 0.30600kg
Product Code. 12980881
Quantity Price
1 £ 49.3 / Pack of 100
Estimated Shipment
22-07-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Closure Color Blue
Material (Liner) Silicone/PTFE
Color Amber
Diameter (Metric) Outer 11.6 mm
Material Glass
Volume (Metric) 1.85 mL
No. per Pack 100
Closure Material Polypropylene
Closure Type Screw Cap
Height (Metric) 32 mm
Closure Size 9-425
Marking Spot Yes
  • ND9
  • 9 mm
  • Glass
  • Amber
  • Patched
  • 1.5 mL
  • Screw Cap
  • Center hole (6 mm)
  • Polypropylene
  • PTFE/Silicone
  • 1.0 mm thickness
  • Reusable two compartment trays protect vials and closure while keeping matching supplies together