Code:
33
Brannan™ LO-tox™ Lab Thermometer
LO-tox™ lab thermometer without calibration certificate.
Additional Details:
Weight:
1.00000kg
Specification
For Use With (Application) | General use |
---|---|
Immersion Type | Total Immersion |
Includes | Brannan non roll end cap |
Length (Metric) | 405 mm |
Material | Glass |
Thermometer Fill | Blue Spirit |
Product Type | Laboratory Thermometer |
Max. Temperature (English) | 123.8°F |
Max. Temperature (Metric) | 51°C |
Min. Temperature (English) | 30.2°F |
Min. Temperature (Metric) | -1°C |
Temperature Range (Metric) | -1°C to +51°C |
For Use With (Equipment) | General Use |
Accuracy | ±0.5°C |
Subdivision | 0.2°C |