Code:
X2
Merck Amicon™ Ultra-2 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
Additional Details:
Weight:
1.00000kg
Includes:
Two microcentrifuge tubes
Specification
Disposable | Single-use |
---|---|
Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
MWCO | 50 kDa |
Sterility | Non-sterile |
Filtration Area | 1 cm2 |
For Use With (Application) | Protein purification, protein concentration and desalting |
Housing Material | Regenerated Cellulose (RC) |
Includes | Two microcentrifuge tubes |
Quantity | 24Pack |
Type | Centrifugal Filters |
Volume (Metric) | 2 mL |
Length (Metric) | 119.7 mm |
Diameter (Metric) | 15.9 mm |