Use promo code "24353" to get your promotional price.
Thermo Scientific™ HisPur™ Ni-NTA Superflow Agarose
Use promo code "24353" to get your promotional price.
Specification
Capacity (Metric) | 20 mg His-Tagged GFP Binding per mL Resin |
---|---|
Content And Storage | Store at 4°C |
Form | Liquid Suspension |
Product Line | HisPur™ |
Quantity | 50 mL |
Target | His-Tagged Protein |
Type | Agarose |
Stationary Phase | Divalent Nickel, Metal Chelate |
Format | Bottle |
Column Type | Affinity, Agarose Resin |
[HisPur Ni-NTA Superflow Agarose, 50 mL.]
[Danger]
[{code=FLAM. LIQ. 3, sequence=1, description=Flammable liquid Category 3}, {code=SKIN SENS. 1, sequence=2, description=Skin sensitiser Category 1}, {code=RESP. SENS. 1, sequence=3, description=Respiratory sensitiser Category 1}, {code=CARC. 1, sequence=4, description=Carcinogenicity Category 1}, {code=REPR. 1B, sequence=5, description=Reproductive toxicity Category 1B}, {code=STOT RE 2, sequence=6, description=Specific target organ toxicity after repeated exposure Category 2}, {code=AQUA CHRONIC 3, sequence=7, description=LONG-TERM AQUATIC HAZARD Chronic 3}]
[{code=H226, sequence=1, description=Flammable liquid and vapour.}, {code=H317, sequence=2, description=May cause an allergic skin reaction.}, {code=H334, sequence=3, description=May cause allergy or asthma symptoms or breathing difficulties if inhaled.}, {code=H350, sequence=4, description=May cause cancer.}, {code=H360D, sequence=5, description=May damage the unborn child.}, {code=H373, sequence=6, description=May cause damage to organs.}, {code=H412, sequence=7, description=Harmful to aquatic life with long lasting effects.}]
[{code=P201, sequence=51, description=Obtain special instructions before use.}, {code=P260, sequence=52, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P302+P352, sequence=53, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P303+P361+P353, sequence=55, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P370+P378, sequence=56, description=In case of fire: Use to extinguish.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Ethyl alcohol, Nickel(II) sulfate hexahydrate(1:1:6)}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS08, sequence=2, description=HEALTH HAZARD}]
Product Identifier
- HisPur Ni-NTA Superflow Agarose, 50 mL.
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Carcinogenicity Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226- Flammable liquid and vapour.
- H317- May cause an allergic skin reaction.
- H334- May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350- May cause cancer.
- H360D- May damage the unborn child.
- H373- May cause damage to organs.
- H412- Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201- Obtain special instructions before use.
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378- In case of fire: Use to extinguish.
Supplemental Information
- MIXTURE LIST- Contains : Ethyl alcohol, Nickel(II) sulfate hexahydrate(1:1:6)