628.41 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Compatible Tips | PIPETMAN DIAMOND Tips D10, DL10, DL10ST, DFL10ST |
---|---|
Adjustable Tip Spacing | No |
Autoclavable | Autoclavable |
Color | Red |
Dispensing Speed | Manual |
Disposable | Reusable |
For Use With | Pipette Tip 0.5 to 20 μL |
Grip Type | Ergonomic |
Material | Polypropylene |
Model | P8x10L |
Number of Channels | 8 |
Product Line | Pipetman |
Programmability | No |
Sterility | Non-sterile |
Type | Pipette |
Volume (Metric) | 0.5 to 10 μL |
Finger Style | Ergonomic |
Quantity | 1 |
Warranty | 1 Year |
Length (English) | 5.98 in. |
Length (Metric) | 15.2 cm |
Width (English) | 12.83 in. |
Width (Metric) | 32.6 cm |
Height (English) | 1.54 in. |
Height (Metric) | 3.9 cm |
Weight (English) | 0.81 lb. |
Color Code | Red |
Fit for Purpose | Certified Quality |