Use promo code "24135" to get your promotional price.
Sartorius Microsart™ @Filter 100 in Trays
Membrane filters suitable for use in the Microsart @Filter units
124.99 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Code:
NEW
Additional Details:
Weight:
1.00000kg
Specification
Color | White |
---|---|
Disposable | Single-use |
Format | Disc |
Material | Cellulose Nitrate |
Material (Membrane) | Cellulose Nitrate |
Pore Size | 0.5 μm |
Sample Volume | 100 mL |
Sterility | Sterile |
Grid Color | Black |
Type | Filter Tip |
Diameter (Metric) | 47 mm |
-
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysis
Buy Now