SI Analytics TitroLine™ TL 7000-M2/20
Features the highest safety for measuring and calibrating through wireless sensor recognition for ID-electrodes. SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M2/20 possesses general titration feautres but offers improved performace.
Code:
NEW
Additional Details:
Weight:
8.00000kg
Specification
Type | TitroLine 7000 Basic Unit |
---|---|
Includes | Brown Glass Bottle For Titrant, GL 45 And S 40-Bottle Adapter, Hoses, Drip Off Tube And Titration Tip. With pH-Combination Electrode IL-pHT-A120MF-DIN-N And Buffer Set |
Material | Polypropylene |
Model | TL 7000-M2/20 |
Temperature Range (Metric) Ambient | 10°C to 40°C |
Volume (Metric) Sample | 20 mL |
Length (Metric) | 45 cm |
Width (Metric) | 15.3 cm |
Depth (Metric) | 29.6 cm |