Use promo code "25339" to get your promotional price.
Thermo Scientific Acros Benzyl chloride, 99.5+%, extra pure, stabilized
Use promo code "25339" to get your promotional price.
Quantity | 10 L |
---|---|
Packaging | Plastic drum |
Chemical Identifiers
CAS | 100-44-7 |
---|---|
MDL Number | MFCD00000889 |
InChI Key | KCXMKQUNVWSEMD-UHFFFAOYSA-N |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
PubChem CID | 7503 |
ChEBI | CHEBI:615597 |
IUPAC Name | chloromethylbenzene |
SMILES | C1=CC=C(C=C1)CCl |
Specification
Chemical Name or Material | Benzyl chloride |
---|---|
Name Note | 99.5+% |
CAS | 100-44-7 |
CAS Min % | 99.5 |
CAS Max % | 100.0 |
CAS | 75-56-9 |
Melting Point | -39.0°C |
Density | 1.1000g/mL |
Boiling Point | 179.0°C |
Flash Point | 67°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.5% min. (GC) |
Packaging | Plastic drum |
Molecular Formula | C7H7Cl |
Linear Formula | C6H5CH2Cl |
Refractive Index | 1.5370 to 1.5390 |
MDL Number | MFCD00000889 |
Quantity | 10 L |
Beilstein | 05,292 |
Merck Index | 15,1132 |
Specific Gravity | 1.1 |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Synonym | benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol |
Solubility Information | Solubility in water: 0.3g/L (20°C). Other solubilities: miscible with ethanol,ethyl ether,chloroform,,acetone,slightly soluble in carbon tetrachloride |
InChI Key | KCXMKQUNVWSEMD-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)CCl |
IUPAC Name | chloromethylbenzene |
Stabilizer | 0.25% propylene oxide |
Molecular Weight (g/mol) | 126.59 |
PubChem CID | 7503 |
ChEBI | CHEBI:615597 |
Viscosity | 1.280 mPa.s (20°C) |
Formula Weight | 126.59 |
Percent Purity | 99.5+% |
Grade | Extra Pure |
Physical Form | Liquid |
Technical documents
[Benzyl chloride, stabilized]
[Danger]
[{code=ACUTE TOX. 3, sequence=1, description=Acute toxicity Category 3}, {code=ACUTE TOX. 4, sequence=2, description=Acute toxicity Category 4}, {code=CARC. 1B, sequence=3, description=Carcinogenicity Category 1B}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}, {code=MET. CORR. 1, sequence=5, description=Substances and mixtures corrosive to metals Category 1}, {code=MUTA. 1B, sequence=6, description=Germ cell mutagenicity Category 1B}, {code=RESP. SENS. 1, sequence=7, description=Respiratory sensitiser Category 1}, {code=SKIN IRRIT. 2, sequence=8, description=Skin corrosion/irritation Category 2}, {code=STOT RE 2, sequence=9, description=Specific target organ toxicity after repeated exposure Category 2}, {code=STOT SE 3, sequence=10, description=Specific target organ toxicity Category 3}]
[{code=H317, sequence=1, description=May cause an allergic skin reaction.}, {code=H318, sequence=2, description=Causes serious eye damage.}, {code=H340, sequence=3, description=May cause genetic defects.}, {code=H350, sequence=4, description=May cause cancer.}, {code=H315, sequence=5, description=Causes skin irritation.}, {code=H373, sequence=6, description=May cause damage to organs.}, {code=H331, sequence=7, description=Toxic if inhaled.}, {code=H335, sequence=8, description=May cause respiratory irritation.}, {code=H302, sequence=9, description=Harmful if swallowed.}, {code=H290, sequence=10, description=May be corrosive to metals.}]
[{code=P280, sequence=50, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=51, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P302+P352, sequence=52, description=IF ON SKIN: Wash with plenty of water/.}, {code=P304+P340, sequence=53, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=54, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=55, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- Benzyl chloride, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Germ cell mutagenicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317- May cause an allergic skin reaction.
- H318- Causes serious eye damage.
- H340- May cause genetic defects.
- H350- May cause cancer.
- H315- Causes skin irritation.
- H373- May cause damage to organs.
- H331- Toxic if inhaled.
- H335- May cause respiratory irritation.
- H302- Harmful if swallowed.
- H290- May be corrosive to metals.
Precautionary Statement
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352- IF ON SKIN: Wash with plenty of water/.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.