Use promo code "24127" to get your promotional price.
Thermo Scientific Acros Hydrochloric acid, 37%, for analysis, (max. 0.000001% Hg)
Use promo code "24127" to get your promotional price.
Quantity | 2.5 L |
---|---|
Packaging | Glass Bottle |
Specification
Chemical Name or Material | Hydrochloric Acid |
---|---|
CAS | 7647-01-0 |
Melting Point | -35.0°C |
Boiling Point | 57.0°C |
Density | 1.1800g/mL |
CAS | 7732-18-5 |
Packaging | Glass Bottle |
Specific Gravity | 1.18 |
Assay Percent Range | 37% |
Quantity | 2.5 L |
Molecular Formula | ClH |
Viscosity | 1.9 mPa.s (15°C) |
Linear Formula | HCl |
MDL Number | MFCD00011324 MFCD00792839 |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Synonym | hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof |
Solubility Information | Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
SMILES | Cl |
IUPAC Name | chlorane |
Molecular Weight (g/mol) | 36.46 |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
Formula Weight | 36.46 |
Percent Purity | 37% |
Grade | Analytical |
Technical documents
[Danger]
[{code=EYE DAM. 1, sequence=1, description=Serious eye damage/eye irritation Category 1}, {code=MET. CORR. 1, sequence=2, description=Substances and mixtures corrosive to metals Category 1}, {code=SKIN CORR. 1B, sequence=3, description=Skin corrosion/irritation Category 1 B}, {code=STOT SE 3, sequence=4, description=Specific target organ toxicity Category 3}]
[{code=H290, sequence=1, description=May be corrosive to metals.}, {code=H314, sequence=2, description=Causes severe skin burns and eye damage.}, {code=H318, sequence=3, description=Causes serious eye damage.}, {code=H335, sequence=4, description=May cause respiratory irritation.}]
[{code=P234, sequence=51, description=Keep only in original container.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=53, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P304+P340, sequence=54, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS05, sequence=1, description=CORROSION}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H290- May be corrosive to metals.
- H314- Causes severe skin burns and eye damage.
- H318- Causes serious eye damage.
- H335- May cause respiratory irritation.
Precautionary Statement
- P234- Keep only in original container.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
-
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.
Buy Now