184.67 GBP valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Description | Nunclon Sphera 96U-well plate, case of 8 |
---|---|
Material | Polystyrene |
Volume (Metric) Well | 300 μL |
Type | Microplate |
Quantity | 8/Cs. |
No. per Pack | 1 Microplate |
No. per Case | 8 Microplates |
Color | Clear |
Surface Treatment | Nunclon Sphera-Treated |
Sterility | Sterile |
No. of Wells | 96 |
Certifications/Compliance | Animal origin-free; nonpyrogenic, endotoxin <0.5 EU/mL; USP biological reactivity class VI; SAL 10-6; ANSI/SBS 1-2004 |
Array | 8 x 12 |
Product Line | Nunc™ |
Packaging | 1/Pack, 8/Case |
Well Shape | U |