1346.63 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Use promo code "25721" to get your promotional price.
Capacity (Metric) | 130 L |
---|---|
Temperature Range | 1°C to 15°C |
Refrigerant | R600A |
Door Material | Double-glazed |
Cabinet Material | Steel |
Interior | White Polystyrene interior containers |
Color | White |
Model | FKUV1613X |
Dimensions (L x W x H) Exterior | 83 H x 60 W x 61.5 D cm |
Depth (Metric) Interior | 43.5 cm |
Height (Metric) Interior | 67 cm |
Width (Metric) Interior | 44 cm |
Voltage | 220/240 V |
Electrical Requirements | 220 - 240 V 50 Hz 120 w |
Frequency | 50 Hz |
Shipping Weight (Metric) | 42 kg |